(±)-3,3-Dimethyl-2-butylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11530-06
A11530-14
CAAAA11530-06EA
341.28
CAD
CAAAA11530-06
CAAAA11530-14
(±)-3,3-Dimethyl-2-butylamine 98%
(±)-3,3-Dimethyl-2-butylamine
Purity: 98%
CAS No.: 3850-30-4
Molecular Formula: C6H15N
Formula Weight: 101.19
UN No.: 2733
MDL No.: MFCD00008078
Formula:
C₆H₁₅N Density: 0.755 Storage Temperature: Ambient |
MDL Number:
MFCD00008078 CAS Number: 3850-30-4 EINECS: 223-353-4 UN: 3286 ADR: "3",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...