Thymidine (2'-deoxythymidine) 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine

A11493-06 A11493-14 A11493-22
CAAAA11493-06EA 121.98 CAD
CAAAA11493-06 CAAAA11493-14 CAAAA11493-22
Thymidine (2'-deoxythymidine) 99%
Thymidine (2'-Deoxythymidine)

Purity: 99% CAS No.: 50-89-5 Molecular Formula: C10H14N2O5 Formula Weight: 242.23 Physical Form: Powder Appearance: White Odor: Odorless Melting Point: 186-190° Beilstein Reference: 89285 Merck Reference: 14,9397 Optical Rotation: +19° (c=1 in water) MDL No.: MFCD00006537

Formula: C₁₀H₁₄N₂O₅
Melting Pt: 186…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00006537
CAS Number: 50-89-5
EINECS: 200-070-4
Merck Index: 14,9397

Order Now

Specification Test Results

Optical rotation +19° (c=1 in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR