Boron trifluoride acetic acid complex (1:2) (35 - 36.5% BF₃)
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11471-AK
A11471-AU
CAAAA11471-AKEA
93.54
CAD
CAAAA11471-AK
CAAAA11471-AU
Boron trifluoride acetic acid complex (1:2) (35 - 36.5% BF₃)
Boron trifluoride acetic acid complex (1:2)
Fieser: 1,69 10,51
Purity: 96%
CAS No.: 373-61-5
Molecular Formula: BF3.2CH3CO2H
Formula Weight: 187.91
Flash Point: 83°(181°F)
Refractive Index: 1.3670
Odor: Pungent
pH:
Formula:
C₄H₈BF₃O₄ Boiling Pt: 143…146 °C Melting Pt: –47 °C Density: 1.353 Flash Pt: 83 °C(181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00036359 CAS Number: 373-61-5 EINECS: 206-768-5 UN: 1742 ADR: "8",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...