4-Nitrocinnamaldehyde predominantly trans 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(4-Nitrophenyl)acrylaldehyde

A11467-18 A11467-06 A11467-14
CAAAAA11467-18EA 1257.68 CAD
CAAAAA11467-18 CAAAA11467-06 CAAAA11467-14
4-Nitrocinnamaldehyde predominantly trans 98%
4-Nitrocinnamaldehyde

Purity: 98% CAS No.: 1734-79-8 Molecular Formula: C9H7NO3 Formula Weight: 177.16 Odor: Characteristic Melting Point: 138-142° Beilstein Reference: 1565424 MDL No.: MFCD00007379

Formula: C₉H₇NO₃
MW: 177.16 g/mol
Melting Pt: 138…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00007379
CAS Number: 1734-79-8
EINECS: 217-076-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR