2-Amino-5-mercapto-1,3,4-thiadiazole 98%

Supplier: Thermo Scientific Chemicals

A11451-14 A11451-22
CAAAA11451-14EA 70.31 CAD
CAAAA11451-14 CAAAA11451-22
2-Amino-5-mercapto-1,3,4-thiadiazole 98%
2-Amino-5-mercapto-1,3,4-thiadiazole

Purity: 98+% CAS No.: 2349-67-9 Molecular Formula: C2H3N3S2 Formula Weight: 133.19 Melting Point: ca 243° dec. Beilstein Reference: 81728

Formula: C₂H₃N₃S₂
Melting Pt: ca 243 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003108
CAS Number: 2349-67-9
EINECS: 219-078-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR