2-(2-Bromoethyl)-1,3-dioxolane 95% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with silver
A11441-09
A11441-18
CAAAA11441-09EA
82.79
CAD
CAAAA11441-09
CAAAA11441-18
2-(2-Bromoethyl)-1,3-dioxolane 95% stabilized
2-(2-Bromoethyl)-1,3-dioxolane
Fieser: 7,37
Purity: 95%
CAS No.: 18742-02-4
Molecular Formula: C5H9BrO2
Formula Weight: 181.04
Storage Temperature: 2°C to 8°C
Flash Point: 65°(149°F)
Refractive Index: 1.4790
Boiling Point: 68-70°/8mm
Beilstein Reference: 103516
UN No.: 2810
MDL No.: MFCD00003216
Formula:
C₅H₉BrO₂ MW: 181.03 g/mol Boiling Pt: 68…70 °C/8 mm Density: 1.512 Flash Pt: 65 °C(149 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003216 CAS Number: 18742-02-4 EINECS: 242-551-1 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...