Dimethyl acetylenedicarboxylate 98%

Supplier: Thermo Scientific Chemicals
Danger

A11437-36 A11437-14 A11437-22
CAAAAA11437-36EA 1933.92 CAD
CAAAAA11437-36 CAAAA11437-14 CAAAA11437-22
Dimethyl acetylenedicarboxylate 98%
Dimethyl acetylenedicarboxylate

Purity: 98% CAS No.: 762-42-5 Molecular Formula: C6H6O4 Formula Weight: 142.11 Flash Point: 86°(186°F) Refractive Index: 1.4480 Melting Point: -18° Boiling Point: 95-98°/19mm Beilstein Reference: 607063 UN No.: 1760 MDL No.: MFCD00008456

Formula: C₆H₆O₄
Boiling Pt: 95…98 °C/19 mm
Melting Pt: –18 °C
Density: 1.156
Flash Pt: 86 °C(186 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008456
CAS Number: 762-42-5
EINECS: 212-098-4
UN: 3265
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR