3-Methylbenzo[b]thiophene-2-carboxaldehyde 98%
Supplier: Thermo Scientific Chemicals
A11375-03
A11375-06
CAAAAA11375-14EA
2021.66
CAD
CAAAAA11375-14
CAAAA11375-03
CAAAA11375-06
3-Methylbenzo[b]thiophene-2-carboxaldehyde 98%
3-Methylbenzo[b]thiophene-2-carboxaldehyde
Purity: 98%
CAS No.: 22053-74-3
Molecular Formula: C10H8OS
Formula Weight: 176.24
Melting Point: 88-90°
Beilstein Reference: 121582
Formula:
C₁₀H₈OS Melting Pt: 88…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052307 CAS Number: 22053-74-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...