Guanosine ≥98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

A11328-14 A11328-22 A11328-18 A11328-30
CAAAAA11328-14EA 90.06 CAD
CAAAAA11328-14 CAAAAA11328-22 CAAAA11328-18 CAAAA11328-30
Guanosine ≥98%
Guanosine

Purity: 98+% CAS No.: 118-00-3 Molecular Formula: C10H13N5O5 Formula Weight: 283.24 Melting Point: ca 250° dec. Beilstein Reference: 625911 Merck Reference: 14,4566 Optical Rotation: -72° (c=1.5 in 0.1N NaOH) MDL No.: MFCD00010182

Formula: C₁₀H₁₃N₅O₅
MW: 283.24 g/mol
Melting Pt: ∼ 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010182
CAS Number: 118-00-3
EINECS: 204-227-8
UN: 2811
ADR: 6.1,III
Merck Index: 14,4566

Order Now

Specification Test Results

Optical rotation -72° (c=1.5 in 0.1N NaOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR