Disodium fumarate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11276-0E
A11276-22
A11276-36
CAAAA11276-0EEA
756.82
CAD
CAAAA11276-0E
CAAAA11276-22
CAAAA11276-36
Disodium fumarate 98%
Disodium fumarate
Purity: 98%
CAS No.: 17013-01-3
Molecular Formula: C4H2Na2O4
Formula Weight: 160.04
Physical Form: Powder
Sensitivity: Hygroscopic
MDL No.: MFCD00064567
Formula:
C₄H₂Na₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00064567 CAS Number: 17013-01-3 EINECS: 241-087-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...