Diisoamyl ether (mixture of isomers) 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11231-AC
A11231-AE
A11231-AP
CAAAA11231-ACEA
46.14
CAD
CAAAA11231-AC
CAAAA11231-AE
CAAAA11231-AP
Diisoamyl ether (mixture of isomers) 96%
Diisoamyl ether (mixture of isomers)
Purity: 96%
CAS No.: 544-01-4
Molecular Formula: [(CH3)2CHCH2CH2]2O
Formula Weight: 158.29
Flash Point: 46°(115°F)
Refractive Index: 1.407
Boiling Point: 172-174°
Beilstein Reference: 1698014
Merck Reference: 14,5118
UN No.: 3271
MDL No.: MFCD00008947
Formula:
C₁₀H₂₂O Boiling Pt: 172…174 °C Density: 0.78 Flash Pt: 46 °C(115 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008947 CAS Number: 544-01-4 EINECS: 208-857-4 UN: 1993 ADR: "3",III Merck Index: 14,5118 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...