2'-Deoxyadenosine monohydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

A11166-14 A11166-03 A11166-06
CAAAAA11166-14EA 769.46 CAD
CAAAAA11166-14 CAAAA11166-03 CAAAA11166-06
2'-Deoxyadenosine monohydrate 99%
2'-Deoxyadenosine monohydrate

Purity: 99% CAS No.: 16373-93-6 Molecular Formula: C10H13N5O3.H2O Formula Weight: 269.26 (251.24anhy) Storage Temperature: 2°C to 8°C Melting Point: 186-189° Beilstein Reference: 5191174 Optical Rotation: -25° (c=0.5 water) MDL No.: MFCD00149364

Formula: C₁₀H₁₃N₅O₃·1H₂O
Melting Pt: 186…189 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149364
CAS Number: 16373-93-6
EINECS: 213-488-7

Order Now

Specification Test Results

Optical rotation -25° (c=0.5 in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR