Formamidine acetate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11158-22
A11158-36
CAAAA11158-22EA
95.43
CAD
CAAAA11158-22
CAAAA11158-36
Formamidine acetate 99%
Formamidine acetate
Fieser: 1,403 2,202 3,147
Purity: 99%
CAS No.: 3473-63-0
Molecular Formula: C3H8N2O2
Formula Weight: 104.11
pH: 8
Melting Point: ca 158° dec.
Beilstein Reference: 3563488
MDL No.: MFCD00012866
Formula:
C₃H₈N₂O₂ Melting Pt: ca 158 °C (dec.) Density: 1.124 Storage Temperature: Ambient |
MDL Number:
MFCD00012866 CAS Number: 3473-63-0 EINECS: 222-442-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...