Carbohydrazide 97%

Supplier: Thermo Scientific Chemicals
Warning

A11145-14 A11145-22 A11145-36
CAAAA11145-14EA 85.95 CAD
CAAAA11145-14 CAAAA11145-22 CAAAA11145-36
Carbohydrazide 97%
Carbohydrazide

Purity: 97% CAS No.: 497-18-7 Molecular Formula: CH6N4O Formula Weight: 90.09 Storage Temperature: 2°C to 8°C Melting Point: 153-158° UN No.: 3077 MDL No.: MFCD00007591

Formula: CH₆N₄O
Melting Pt: 153…158 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007591
CAS Number: 497-18-7
EINECS: 207-837-2
UN: 3077
ADR: "9",III
Merck Index: 14,1804

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR