S-Methylisothiouronium sulfate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
S-Methylisothiourea sulphate, 2-Methyl-2-thiopseudourea sulphate, S-Methylthiouronium sulfate, S-Methylthiouronium sulphate, 2-Methyl-2-thiopseudourea hemisulphate, S-Methylisothiourea sulfate, 2-Methyl-2-thiopseudourea sulfate, 2-Methyl-2-thiopseudourea hemisulfate
A11044-30
A11044-0E
A11044-22
A11044-36
CAAAAA11044-30EA
73.63
CAD
CAAAAA11044-30
CAAAA11044-0E
CAAAA11044-22
CAAAA11044-36
S-Methylisothiouronium sulfate 98+%
S-Methylisothiouronium sulphate
Purity: 98+%
CAS No.: 867-44-7
Molecular Formula: C4H12N4S2.H2SO4
Formula Weight: 278.38
Melting Point: ca 235° dec.
Beilstein Reference: 3917217
Formula:
C₄H₁₂N₄S₂·H₂SO₄ MW: 278.38 g/mol Melting Pt: ∼ 235 °C (dec.) Density: 1.28 Storage Temperature: Ambient |
MDL Number:
MFCD00013055 CAS Number: 867-44-7 EINECS: 212-759-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...