3-Nitro-L-tyrosine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-Tyr(3-NO2)-OH, 3-Nitro-L-tyrosine

A11018-06 A11018-14 A11018-22
CAAAA11018-06EA 74.42 CAD
CAAAA11018-06 CAAAA11018-14 CAAAA11018-22
3-Nitro-L-tyrosine 98%
3-Nitro-L-tyrosine

Purity: 98% CAS No.: 621-44-3 Molecular Formula: C9H10N2O5 Formula Weight: 226.19 Melting Point: ca 230° dec. Beilstein Reference: 2813157

Formula: C₉H₁₀N₂O₅
MW: 226.3 g/mol
Melting Pt: ca 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007123
CAS Number: 621-44-3
EINECS: 210-688-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR