(±)-Ethyl-2-bromopropionate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: DL-Ethyl 2-bromopropionate, Ethyl 2-bromopropionate

A10820-18 A10820-30 A10820-36
CAAAAA10820-18EA 42.66 CAD
CAAAAA10820-18 CAAAAA10820-30 CAAAA10820-36
(±)-Ethyl-2-bromopropionate 98+%
(±)-Ethyl 2-bromopropionate

Purity: 98+% CAS No.: 535-11-5 Molecular Formula: C5H9BrO2 Formula Weight: 181.03 Flash Point: 51°(123°F) Refractive Index: 1.4460 Boiling Point: 158-160° Beilstein Reference: 773920 Merck Reference: 14,3772 UN No.: 1992 MDL No.: MFCD00000144

Formula: C₅H₉BrO₂
MW: 181.03 g/mol
Boiling Pt: 158…160 °C
Density: 1.391
Flash Pt: 51 °C(123 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000144
CAS Number: 535-11-5
EINECS: 208-609-5
UN: 2920
ADR: "8",II
Merck Index: 14,3772

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR