Adenosine 99%
Supplier: Thermo Scientific Chemicals
A10781-09
A10781-18
A10781-30
CAAAA10781-09EA
70.94
CAD
CAAAA10781-09
CAAAA10781-18
CAAAA10781-30
Adenosine 99%
Adenosine
Purity: 99%
CAS No.: 58-61-7
Molecular Formula: C10H13N5O4
Formula Weight: 267.25
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Odor: Odorless
Melting Point: 234-237°
Beilstein Reference: 93029
Merck Reference: 14,153
Optical Rotation: -70° (c=2 in 5% NaOH)
MDL No.: MFCD00005752
Formula:
C₁₀H₁₃N₅O₄ Melting Pt: 234…237 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005752 CAS Number: 58-61-7 EINECS: 200-389-9 Merck Index: 14,153 |
Specification Test Results
Optical rotation | -70° (c=2 in 5% NaOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...