3-Thienylacetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10700-06
A10700-14
A10700-22
CAAAA10700-06EA
77.58
CAD
CAAAA10700-06
CAAAA10700-14
CAAAA10700-22
3-Thienylacetic acid 98%
Purity: 98%
CAS No.: 6964-21-2
Molecular Formula: C6H6O2S
Formula Weight: 142.18
Melting Point: 77-81°
Boiling Point: 129°/2.5mm
UN No.: 3261
MDL No.: MFCD00005473
Formula:
C₆H₆O₂S Boiling Pt: 129 °C/2.5 mm Melting Pt: 77…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005473 CAS Number: 6964-21-2 EINECS: 230-166-1 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...