Diethyl benzylphosphonate 99%

Supplier: Thermo Scientific Chemicals
Warning

A10645-0B A10645-18 A10645-30
CAAAA10645-0BEA 2292.58 CAD
CAAAA10645-0B CAAAA10645-18 CAAAA10645-30
Diethyl benzylphosphonate 99%
Diethyl benzylphosphonate

Purity: 99% CAS No.: 1080-32-6 Molecular Formula: C11H17O3P Formula Weight: 228.23 Flash Point: 110°(230°F) Refractive Index: 1.4975 Boiling Point: 106-108°/1mm Beilstein Reference: 2580931 MDL No.: MFCD00009078

Formula: C₁₁H₁₇O₃P
Boiling Pt: 106…108 °C/1 mm
Density: 1.094
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009078
CAS Number: 1080-32-6
EINECS: 214-097-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR