Tetrabutylammonium fluoride 1 M in THF

Supplier: Thermo Scientific Chemicals
Danger

A10588-AC A10588-AE A10588-AP
CAAAAA10588-ACEA 78.37 CAD
CAAAAA10588-AC CAAAAA10588-AE CAAAAA10588-AP
Tetrabutylammonium fluoride 1 M in THF
Tetrabutylammonium fluoride

Fieser: 4,477 14,293 15,304 17,324 18,344 19,319 20,357 21,407


CAS No.: 429-41-4 Molecular Formula: C16H36FN Formula Weight: 261.47 Flash Point: -17°(1°F) Beilstein Reference: 3570522 UN No.: 2924 MDL No.: MFCD00011747

Formula: C₁₆H₃₆FN
MW: 261.47 g/mol
Density: 0.903
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011747
CAS Number: 429-41-4
EINECS: 207-057-2
UN: 2924
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR