Glycylglycine (Gly-Gly)
Supplier: Thermo Scientific Chemicals
A10523-36
A10523-14
A10523-22
CAAAAA10523-36EA
739.44
CAD
CAAAAA10523-36
CAAAA10523-14
CAAAA10523-22
Glycylglycine (Gly-Gly)
Proteins and Peptides
Buffer with a useful pH range 7.5 to 8.9
- EINECS: 209-127-8
- GHS symbol and signal word: GHS07 and warning
- Melting point: ca 220 °C (dec.)
- Merck index: 14,4503
Purity: 99%
CAS No.: 556-50-3
Molecular Formula: H2NCH2CONHCH2CO2H
Formula Weight: 132.12
Melting Point: ca 220° dec.
Beilstein Reference: 1765223
Merck Reference: 14,4503
MDL No.: MFCD00008130
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...