Aminoacetaldehyde diethyl acetal 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,2-Diethoxyethanamine, 2-Aminoacetaldehyde diethylacetal
A10427-14
A10427-22
CAAAA10427-14EA
103.33
CAD
CAAAA10427-14
CAAAA10427-22
Aminoacetaldehyde diethyl acetal 98%
Aminoacetaldehyde diethyl acetal
Purity: 98%
CAS No.: 645-36-3
Molecular Formula: C6H15NO2
Formula Weight: 133.19
Flash Point: 45°(113°F)
Refractive Index: 1.4170
Boiling Point: 162-163°
Beilstein Reference: 506067
UN No.: 2733
MDL No.: MFCD00008136
Formula:
C₆H₁₅NO₂ Boiling Pt: 162…163 °C Density: 0.916 Flash Pt: 45 °C(113 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008136 CAS Number: 645-36-3 EINECS: 211-439-4 UN: 2733 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...