2-Thienylacetic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Thiopheneacetic acid, Thiophene-2-acetic acid, 2-(Thiophen-2-yl)acetic acid

A10394-14 A10394-22 A10394-36
CAAAA10394-14EA 84.06 CAD
CAAAA10394-14 CAAAA10394-22 CAAAA10394-36
2-Thienylacetic acid 98%
2-Thienylacetic acid

Purity: 98% CAS No.: 1918-77-0 Molecular Formula: C6H6O2S Formula Weight: 142.18 Melting Point: 61-65° Boiling Point: 160°/22mm UN No.: 3261 MDL No.: MFCD00005458

Formula: C₆H₆O₂S
MW: 142.18 g/mol
Boiling Pt: 160 °C (22 mmHg)
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00005458
CAS Number: 1918-77-0
EINECS: 217-639-8
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR