Dilithium tetraborate trihydrate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lithium tetraborate trihydrate

A10358-22 A10358-36
CAAAAA10358-22EA 76.16 CAD
CAAAAA10358-22 CAAAAA10358-36
Dilithium tetraborate trihydrate 96%
di-Lithium tetraborate trihydrate

Purity: 96% CAS No.: 37195-62-3 Molecular Formula: Li2B4O7.3H2O Formula Weight: 223.17 (169.12anhy) Merck Reference: 14,5524 MDL No.: MFCD00167253

Formula: B₄Li₂O₇·3H₂O
MW: 223.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00167253
CAS Number: 37195-62-3
EINECS: 234-514-3
Merck Index: 14,5524

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR