tert-Butyl-N-hydroxycarbamate 98+%
Supplier: Thermo Scientific Chemicals
A10349-03
A10349-06
A10349-14
CAAAA10349-03EA
45.35
CAD
CAAAA10349-03
CAAAA10349-06
CAAAA10349-14
tert-Butyl-N-hydroxycarbamate 98+%
tert-Butyl N-hydroxycarbamate
Purity: 98+%
CAS No.: 36016-38-3
Molecular Formula: (CH3)3COCONHOH
Formula Weight: 133.15
Storage Temperature: 2°C to 8°C
Melting Point: 55-58°
Beilstein Reference: 1756546
MDL No.: MFCD00002107
Formula:
C₅H₁₁NO₃ Melting Pt: 55…58 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002107 CAS Number: 36016-38-3 EINECS: 252-836-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...