Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Gly-OEt.HCl, Glycine Ethyl Ester HCl, Ethyl Glycinate Hydrochloride

A10315-0E A10315-22 A10315-36
CAAAA10315-0EEA 475.58 CAD
CAAAA10315-0E CAAAA10315-22 CAAAA10315-36
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl) 99%
Glycine ethyl ester hydrochloride (H-Gly-OEt.HCl)

Purity: 99% CAS No.: 623-33-6 Molecular Formula: CH3CH2O2CCH2NH2.HCl Formula Weight: 139.58 Melting Point: 144-148° Beilstein Reference: 3594034

Formula: C₄H₁₀ClNO₂
MW: 139.58 g/mol
Melting Pt: 144…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00012871
CAS Number: 623-33-6
EINECS: 210-787-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR