Cytidine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

A10261-09 A10261-18 A10261-30
CAAAA10261-09EA 112.02 CAD
CAAAA10261-09 CAAAA10261-18 CAAAA10261-30
Cytidine 99%
Cytidine

Purity: 99% CAS No.: 65-46-3 Molecular Formula: C9H13N3O5 Formula Weight: 243.22 Melting Point: ca 215° dec. Beilstein Reference: 89173 Merck Reference: 14,2786 Optical Rotation: +29° (c=9 in water) MDL No.: MFCD00006545

Formula: C₉H₁₃N₃O₅
MW: 243.22 g/mol
Melting Pt: ca 215 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006545
CAS Number: 65-46-3
EINECS: 200-610-9
Merck Index: 14,2786

Order Now

Specification Test Results

Optical rotation +29° (c=9 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR