4-Ethoxymethylene-2-phenyl-2-oxazoline-5-one 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
(4E)-4-(Ethoxymethylene)-2-phenyl-1,3-oxazol-5(4H)-one, 4-Ethoxymethylene-2-phenyloxazolin-5-one
A10155-03
A10155-06
CAAAA10155-03EA
69.84
CAD
CAAAA10155-03
CAAAA10155-06
CAAAA10155-14
4-Ethoxymethylene-2-phenyl-2-oxazoline-5-one 98+%
4-Ethoxymethylene-2-phenyl-2-oxazoline-5-one
Purity: 98+%
CAS No.: 15646-46-5
Molecular Formula: C12H11NO3
Formula Weight: 217.22
Storage Temperature: 2°C to 8°C
Melting Point: 93-97°
Beilstein Reference: 163055
MDL No.: MFCD00003204
Formula:
C₁₂H₁₁NO₃ Melting Pt: 93…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD03225480 CAS Number: 15646-46-5 EINECS: 239-713-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...