Pentaethylene glycol 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,6,9,12-Tetraoxatetradecane-1,14-diol

A10133-09 A10133-18 A10133-30
CAAAAA10133-09EA 205.4 CAD
CAAAAA10133-09 CAAAAA10133-18 CAAAAA10133-30
Pentaethylene glycol 98+%
Pentaethylene glycol

Purity: 98+% CAS No.: 4792-15-8 Molecular Formula: H(OCH2CH2)5OH Formula Weight: 238.28 Flash Point: 110°(230°F) Refractive Index: 1.4620 Boiling Point: 338-340° Beilstein Reference: 1635593 MDL No.: MFCD00002878

Formula: C₁₀H₂₂O₆
MW: 238.28 g/mol
Boiling Pt: 338…340 °C
Density: 1.126
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002878
CAS Number: 4792-15-8
EINECS: 225-341-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR