Lutetium, AAS Standard Solution, Specpure®, Lu 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
89886-AE
89886-AP
CAAA89886-AEEA
104.44
CAD
CAAA89886-AE
CAAA89886-AP
Lutetium, AAS Standard Solution, Specpure®, Lu 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Lutetium Standards
Molecular Formula: Lu2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011098
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...