Lanthanum, compound with nickel (1:5) ≥99.5% (metals basis), powder -100 mesh

Supplier: Thermo Scientific Chemicals

89133-06 89133-14
CAAA89133-06EA 137.78 CAD
CAAA89133-06 CAAA89133-14
Lanthanum, compound with nickel (1:5) ≥99.5% (metals basis), powder -100 mesh
Lanthanum, compound with nickel (1:5)

Purity: 99.5% CAS No.: 12196-72-4 Molecular Formula: LaNi5 Physical Form: Intermetallic, Packaged under argon Sensitivity: Air and Moisture Sensitive Odor: Odorless UN No.: 3178 MDL No.: MFCD00198177

Formula: LaNi₅
MDL Number: MFCD00198177
CAS Number: 12196-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR