Ruthenium, AAS Standard Solution, Specpure®, Ru 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
89117-AE
89117-AP
CAAA89117-AEEA
92.59
CAD
CAAA89117-AE
CAAA89117-AP
Ruthenium, AAS Standard Solution, Specpure®, Ru 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Ruthenium Standards
Molecular Formula: RuCl3 in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011207
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...