Zinc, AAS Standard Solution, Specpure®, Zn 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88118-AE
88118-AP
CAAA88118-AEEA
68.89
CAD
CAAA88118-AE
CAAA88118-AP
Zinc, AAS Standard Solution, Specpure®, Zn 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Zinc Standards
Molecular Formula: Zn in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011291
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...