Vanadium, AAS Standard Solution, Specpure®, V 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88116-AE
88116-AP
CAAA88116-AEEA
67.31
CAD
CAAA88116-AE
CAAA88116-AP
Vanadium, AAS Standard Solution, Specpure®, V 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Vanadium Standards
Molecular Formula: V2O5 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011453
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...