Tin, AAS Standard Solution, Specpure®, Sn 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88112-AE
88112-AP
CAAA88112-AEEA
72.21
CAD
CAAA88112-AE
CAAA88112-AP
Tin, AAS Standard Solution, Specpure®, Sn 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Tin Standards
Molecular Formula: Sn in 20% HCl
Physical Form: Liquid
Odor: Characteristic
UN No.: 1789
MDL No.: MFCD00133862
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...