Thallium, AAS Standard Solution, Specpure®, Tl 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88111-AE
88111-AP
CAAA88111-AEEA
62.09
CAD
CAAA88111-AE
CAAA88111-AP
Thallium, AAS Standard Solution, Specpure®, Tl 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Thallium Standards
Molecular Formula: Tl in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134063
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...