Strontium, AAS Standard Solution, Specpure®, Sr 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88098-AE
88098-AP
CAAA88098-AEEA
65.1
CAD
CAAA88098-AE
CAAA88098-AP
Strontium, AAS Standard Solution, Specpure®, Sr 1000µg/ml, Thermo Scientific
Standards
Cationic Standards
Strontium Standards
Molecular Formula: Sr(NO3)2 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134060
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...