Samarium, AAS Standard Solution, Specpure®, Sm 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88092-AE
88092-AP
CAAA88092-AEEA
63.04
CAD
CAAA88092-AE
CAAA88092-AP
Samarium, AAS Standard Solution, Specpure®, Sm 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Samarium Standards
Molecular Formula: Sm in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011233
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...