Rhenium, AAS standard solution, Specpure®, Re 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88089-AE
88089-AP
CAAA88089-AEEA
92.59
CAD
CAAA88089-AE
CAAA88089-AP
Rhenium, AAS standard solution, Specpure®, Re 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Rhenium Standards
Molecular Formula: Re in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011195
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...