Praseodymium, AAS Standard Solution, Specpure®, Pr 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88088-AE
88088-AP
CAAA88088-AEEA
71.26
CAD
CAAA88088-AE
CAAA88088-AP
Praseodymium, AAS Standard Solution, Specpure®, Pr 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Praseodymium Standards
Molecular Formula: Pr6O11 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011174
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...