Platinum, AAS Standard Solution, Specpure®, Pt 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88086-AE
88086-AP
CAAA88086-AEEA
188.02
CAD
CAAA88086-AE
CAAA88086-AP
Platinum, AAS Standard Solution, Specpure®, Pt 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Platinum Standards
Molecular Formula: Pt in 20% HCl
Physical Form: Liquid
Odor: Characteristic
UN No.: 1789
MDL No.: MFCD00011179
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...