Specpure® Niobium, AAS Standard Solution, Nb 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88083-AE
88083-AP
CAAA88083-AEEA
60.36
CAD
CAAA88083-AE
CAAA88083-AP
Specpure® Niobium, AAS Standard Solution, Nb 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Niobium Standards
Molecular Formula: NbCl5 in 2% HF
Physical Form: Liquid
UN No.: 1790
MDL No.: MFCD00011126
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...