Nickel, AAS Standard Solution, Specpure®, Ni 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88082-AE
88082-AP
CAAA88082-AEEA
67.62
CAD
CAAA88082-AE
CAAA88082-AP
Nickel, AAS Standard Solution, Specpure®, Ni 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Nickel Standards
Molecular Formula: Ni in 5% HNO3
Physical Form: Liquid
Appearance: Colorless
UN No.: 3264
MDL No.: MFCD00011137
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...