Manganese, AAS standard solution, Specpure®, Mn 1000µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

88078-AE 88078-AP
CAAA88078-AEEA 67.62 CAD
CAAA88078-AE CAAA88078-AP
Manganese, AAS standard solution, Specpure®, Mn 1000µg/ml, Thermo Scientific
Standards Cationic Standards Manganese Standards

Molecular Formula: Mn in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011111

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR