Magnesium, AAS Standard Solution, Specpure®, Mg 1000
Supplier: Thermo Scientific Chemicals
88077-AE
88077-AP
CAAA88077-AEEA
67.62
CAD
CAAA88077-AE
CAAA88077-AP
Magnesium, AAS Standard Solution, Specpure®, Mg 1000
Standards
Cationic Standards
Magnesium Standards
Molecular Formula: Mg in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085308
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...