Lithium, AAS Standard Solution, Specpure®, Li 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88076-AE
88076-AP
CAAA88076-AEEA
63.36
CAD
CAAA88076-AE
CAAA88076-AP
Lithium, AAS Standard Solution, Specpure®, Li 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Lithium Standards
Molecular Formula: Li2CO3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134051
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...