Lead, AAS Standard Solution, Specpure®, Pb 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88075-AE
88075-AP
CAAA88075-AEEA
76.31
CAD
CAAA88075-AE
CAAA88075-AP
Lead, AAS Standard Solution, Specpure®, Pb 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Lead Standards
Pb in 5% HNO3
UN No.: 3264
MDL No.: MFCD00134050
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...