Hafnium, AAS standard solution, Specpure®, Hf 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
88069-AE
88069-AP
CAAA88069-AEEA
66.2
CAD
CAAA88069-AE
CAAA88069-AP
Hafnium, AAS standard solution, Specpure®, Hf 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Hafnium Standards
Molecular Formula: HfCl2O in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011032
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...