Gold, AAS standard solution, Specpure®, Au 1000 µg/ml, Thermo ScientificAesar
Supplier: Thermo Scientific Chemicals
88068-AE
88068-AP
CAAA88068-AEEA
169.06
CAD
CAAA88068-AE
CAAA88068-AP
Gold, AAS standard solution, Specpure®, Au 1000 µg/ml, Thermo ScientificAesar
Standards
Cationic Standards
Gold Standards
Molecular Formula: Au in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00003436
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...